Toluene ≥99.5%, BAKER ANALYZED® ACS, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIALS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

8077.2500EA 0 INR
8077.2500
Toluene ≥99.5%, BAKER ANALYZED® ACS, J.T.Baker®
Toluene
Formula: C₆H₅CH₃
MW: 92.14 g/mol
Boiling Pt: 110.6
Melting Pt: –95 °C
Density: 0.867 g/cm³ (20 °C)
Flash Pt: 4
Storage Temperature: Ambient
MDL Number: MFCD00008512
CAS Number: 108-88-3
EINECS: 203-625-9
UN: 1294
ADR: 3 II
REACH: 05-2114615130-69
Merck Index: 13,09607
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=44efa31292a7dce6d9e60bcbaec208eb42d34e32&vid=7b8a82c36ede4fc1e8822a573f5ce08743569a24&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Exceeds ACS Specifications
Assay (by GC) min. 99.5 %
Benzene max. 0.1 %
Color (APHA) max. 10
Residue after Evaporation max. 0.001 %
Sulfur Compounds (as S) max. 0.003 %
Substances Darkened by H2SO4 passes test
Water (H2O) max. 0.03 %
Trace Impurities (in ppm)
Aluminium (Al) max. 0.5
Boron (B) max. 0.02
Barium (Ba) max. 0.1
Calcium (Ca) max. 0.5
Cadmium (Cd) max. 0.05
Cobalt (Co) max. 0.02
Chromium (Cr) max. 0.02
Copper (Cu) max. 0.02
Iron (Fe) max. 0.1
Magnesium (Mg) max. 0.1
Manganese (Mn) max. 0.02
Nickel (Ni) max. 0.02
Lead (Pb) max. 0.1
Tin (Sn) max. 0.1
Zinc (Zn) max. 0.1


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR