Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

VWR®, Standards for water determination by the Karl Fischer method

Supplier: VWR Chemicals

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:


85482.005EA 58521.11 INR InStock
85482.005 85483.002 85483.005 85483.020 85484.002 85484.005 85484.020 85485.002 85485.005 85485.020 85487.004 85488.230 85489.260 85692.100 85449.100
VWR®, Standards for water determination by the Karl Fischer method
Standards Water Determination (by Karl Fischer method) Standards
The tightening of regulatory requirements and the increasing demands of QA/QC methodology is becoming more important across all market sectors. The demand for standards to support testing methods continues to increase, and this growth applies to water determination according to Karl Fischer. Avantor now offers a complete range of Karl Fischer water standards as CRMs to calibrate and monitor equipment and check test results.

  • CRMs acc. to ISO 17025 / ISO 17034
  • CRM acc. to ASTM (in liquid petroleum)
  • Broad product range for both volumetric + KF coulometric titrations and indirect oven method
  • Liquid and solid water standards available
  • Supplied with a detailed Certificate of Analysis

Our range of Karl Fischer water standards are Certified Reference Materials acc to ISO 17025 and ISO 17034. The standards are available both in liquid and solid form.

We also offer a range of Karl Fischer water standards as CRM for determination in liquid petroleum according to ASTM. The water content in standards in liquid petroleum is determined using the ASTM D6304 method.

Certifications: The water standards in liquid petroleum conform to ASTM D1744, E1064, D4377

Packaging: When the Cat. No. (Pk) ends with a
0.005 then it is 10×5 ml
0.002 then it is 10×2 ml
0.020 then it is 5×20 ml


Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

WRITE A REVIEW

REVIEWS (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

Print These Terms