Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
VWR®, Spreaders, L-Shaped, Disposable
Supplier: VWR BRAND PRIVATE LABEL USA
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
89042-021
89042-021EA 0 INR InStock
89042-021
VWR®, Spreaders, L-Shaped, Disposable
Spreaders Cell Spreaders
PP, sterile, disposable L-shaped cell spreaders are ideal for spreading bacterial cultures evenly.
  • L-shape allows application across the entire Petri dish or plate surface
  • Smooth spreading surface with slight upward return reduces the risk of damaging the agar sample
  • Spreaders are presterilised, eliminating the need for flaming or autoclaving
Packaging: Packaged in resealable bags of 10.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms