VWR®, Fuel testing standards - jet aviation fuel
Supplier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
99122.230EA 0 INR
99122.230 99123.230 99124.230 99126.230 99127.230
VWR®, Fuel testing standards - jet aviation fuel
Standards Petroleum Industry Standards
Fuel testing Certified Reference Materials are for the verification of laboratory instruments used in testing petroleum and derivative products. This includes, but is not limited to, the analysis and measurement of: Acidity content, sulphur content, aniline point, Fluorescent Indicator Adsorption (FIA) aromatics, freezing point.
  • Highest level of accreditation guaranteed, providing the most credible data currently available worldwide
  • Low levels of uncertainty achieves maximum accuracy of data
  • All data generated exclusively by ISO 17025 accredited independent laboratories
  • Ensures good inter-laboratory correlation
  • Certified in accordance with ASTM / EN or IP test method protocols
  • Fully traceable to international standards
  • Tamper-evident security packaging
Certifications: Fully certified in accordance with ISO 17034 under accreditation by UKAS.