Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
BeneSphera™ CF Lyse, 3 Part Differential
Supplier: BENESPHERA
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
15001081
BENP15001081EA 1511.01 INR InStock
BENP15001081
BeneSphera™ CF Lyse, 3 Part Differential
Clinical Diagnostic Supplies Clinical Reagents and Kits
For diluting blood samples in fully automated 3 part analyser.
  • Best fit for 3 part hematology analyser
  • Storage at RT
  • Long shelf life
Caution: Avoid the contact with eyes and skin.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms