Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
BeneSphera™, Malaria Plasmodium vivax/Plasmodium falciparum Rapid Test
Supplier: BENESPHERA
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
15000906 15000907 15001258 15001260
BENP15000906EA 0 INR InStock
BENP15000906 BENP15000907 BENP15001258 BENP15001260
BeneSphera™, Malaria Plasmodium vivax/Plasmodium falciparum Rapid Test
Clinical Diagnostic Supplies Clinical Reagents and Kits
BeneSphera™ Malaria Plasmodium vivax/Plasmodium falciparum Rapid Test is for the differential diagnosis of Plasmodium vivax and Plasmodium falciparum.
  • Detection of pLDH of Plasmodium vivax and HRP-II of Plasmodium falciparum
  • Whole blood specimen
  • Takes 20 to 30 minutes for test result
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms