BeneSphera™ Blood Collection Tubes, Vacuum/Non Vacuum
Supplier: BENESPHERA
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
16008377 16008378 16008379 16008380 16008381 16008382 16008383 16008384 16008385 16008386 16008387 16008388 16008389 16008390 16008391 16008392 16008393 16008394 16008395 16008396 16008397 16008398 16008399 16008400 16008401 16008402 16008403 16008404 16008405 16008406 16008407 16008408 16008409 16008410 16008411 16008412 16008413 16008414 16008415
BENP16008377EA 261 INR
BENP16008377 BENP16008378 BENP16008379 BENP16008380 BENP16008381 BENP16008382 BENP16008383 BENP16008384 BENP16008385 BENP16008386 BENP16008387 BENP16008388 BENP16008389 BENP16008390 BENP16008391 BENP16008392 BENP16008393 BENP16008394 BENP16008395 BENP16008396 BENP16008397 BENP16008398 BENP16008399 BENP16008400 BENP16008401 BENP16008402 BENP16008403 BENP16008404 BENP16008405 BENP16008406 BENP16008407 BENP16008408 BENP16008409 BENP16008410 BENP16008411 BENP16008412 BENP16008413 BENP16008414 BENP16008415
BeneSphera™ Blood Collection Tubes, Vacuum/Non Vacuum
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Blood Sample Collection/Preparation
A complete range for high quality blood collection tubes for the pre-analytical phase of clinical diagnosis.
  • Leak-proof tubes
  • Superior tubes
  • Bromo butyl rubber for unparalleled performance
  • Uniform spray pattern
  • Sterilisation assurance
  • Made from high quality, clear PET/PP
  • Environmentally acceptable, can be recycled or incinerated
These can support mechanical stresses of storage, transport, vacuum filling and sample processing.