Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
Cleanroom wipes, Anticon® Gold StandardWeight™
Supplier: CONTEC
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
495353-931
CONT495353-931EA 0 INR InStock
CONT495353-931
Cleanroom wipes, Anticon® Gold StandardWeight™
Wipes Cleanroom Wipes Dry Cleanroom Wipes
100% no-run interlock knit polyester wipes with sealed edges.
  • Sealed edges reduce particle and fibre generation
  • Lighter basis weight provides superior value for general cleaning applications
  • Low sub-micron particles and fibre counts leaving less behind on surfaces
  • High sorptive capacity and fast adsorption rate for superior spill control
  • Cleanroom laundered and packed in a class 4 cleanroom
Anticon® Gold StandardWeight™ cleanroom wipes are made from 100% continuous filament polyester with a double-knit, no-run interlock construction. The wipes exhibitMore Product Information
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms