Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
E.Z.N.A.® water DNA kit
Supplier: OMEGA BIOTEK
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
D5525-01
D5525-01EA 39805.23 INR InStock
D5525-01
E.Z.N.A.® water DNA kit
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
The E.Z.N.A.® Water DNA kit is formulated to isolate high purity cellular DNA from water samples typically containing humic acid and inhibitors of PCR. This kit has been successfully used to isolate DNA from tough-to-lyse bacteria, fungi, and algae that inhabit a range of samples including tap water, lake, river and sewage samples. Isolated DNA can be used for most downstream applications including PCR, Southern blotting, SNP analysis.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms