Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
MicroElute® Cycle-Pure & Gel Extraction Kit, Omega Bio-tek
Supplier: OMEGA BIOTEK
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
D6294-02 D6294-00
D6294-02EA 0 INR InStock
D6294-02 D6294-00
MicroElute® Cycle-Pure & Gel Extraction Kit, Omega Bio-tek
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
MicroElute® Clean Up system, designed for rapid DNA clean up.
  • Elution volumes – 10-15 µL
  • Generation of highly concentrated DNA
  • 10 µg DNA binding capacity
  • Rapid isolation – less than 15 minutes
The E.Z.N.A.® family of products is an innovative system that radically simplifies the extraction and purification of nucleic acids from a variety of sources. The key to this system is the HiBind® matrix that specifically, but reversibly, binds DNA or RNA under optimized conditions allowing proteins and other contaminants to be removed. Nucleic acids are easily eluted with deionized water or a low salt buffer.
The MicroElute® Clean Up system, designed for rapid DNA clean up, includes:
• MicroElute® Cycle-Pure – for direct purification of double or single stranded PCR products (100 bp – 10 kb) from amplification reactions
• MicroElute® Gel Extraction – for extraction of DNA fragments from standard, or low-melt agarose gels in TAE (Tris-acetate/EDTA) or TBE (Tris-borate/EDTA) buffer
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms