Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Kanamycin sulphate 50 mg/ml in solution, Ultra Pure Grade

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:


E713-20MLEA 4506.75 INR InStock
E713-20ML
Kanamycin sulphate 50 mg/ml in solution, Ultra Pure Grade
Kanamycin sulphate


Formula: C₁₈H₃₈N₄O₁₅S
MW: 582.6 g/mol
MDL Number: MFCD00070253
CAS Number: 25389-94-0
Order Now
Specification Test Results
Clarity PASS / Fail
Specific Gravity (20 °C) 1,0186 - 1,0427

 

ORDER

RELATED INFORMATION

WRITE A REVIEW

REVIEWS (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

Print These Terms