Avantor® ACE® Excel® Glycan, Analytical HPLC/UHPLC Columns
Supplier: HICHROM SELF MANUFACTURED
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
EXL-1116-1002EA 43870 INR
EXL-1116-1002 EXL-1116-1502 EXL-1116-1546 EXL-1716-1002
Avantor® ACE® Excel® Glycan, Analytical HPLC/UHPLC Columns
Chromatography Columns
Avantor® ACE® Glycan is a high performance stationary phase designed for the analysis of 2-aminobenzamide labelled glycans by hydrophilic interaction liquid chromatography (HILIC). These ultra-inert, high efficiency columns provide excellent column lifetime and reproducibility with buffers and mobile phases typically utilised for glycan analysis. Two particle sizes are available to enable both high resolution and rapid separations on HPLC and UHPLC instrumentation.
  • High efficiency 1,7 and 3 µm particle sizes for high resolution glycan separations
  • Ultra-
More Product Information