Sulphuric acid 0.01 mol/l (0.02 N) in aqueous solution 0.01 mol/l (0.02 N) in aqueous solution

Supplier: HACH
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

20353
HACH20353EA 0 INR
HACH20353
Sulphuric acid 0.01 mol/l (0.02 N) in aqueous solution 0.01 mol/l (0.02 N) in aqueous solution
Sulphuric acid
Formula: H₂SO₄
MW: 98.08 g/mol
Boiling Pt: 330
Melting Pt: 10.38
Density: 1.01 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064589
CAS Number: 7664-93-9
EINECS: 231-639-5
UN: 2796
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR