Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
Sulphate test kit, SF-1
Supplier: HACH
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
225100
HACH225100EA 0 INR InStock
HACH225100
Sulphate test kit, SF-1
Environmental Test Kits
High concentrations of magnesium or sodium sulphate in water act as cathartics. This test kit provides economical determination of sulphate levels using the Extinction/Turbidimetric methods.
Delivery information: Includes graduated cylinder, mixing bottle, reagent, dipstick comparator, instructions and carrying case.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms