Fine vacuum ball valves, manual
Supplier: GARDNER DENVER
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
700048 700047
ILMV700048EA 0 INR
ILMV700048 ILMV700047
Fine vacuum ball valves, manual
Valves Vacuum Valves
These fine vacuum ball valves feature a small flange made of nickle plated aluminium.
  • Nickel-plated brass casing, polished brass ball, plastic (graphitised) seals