Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
Ball check valves, manual, ABV series
Supplier: GARDNER DENVER
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
700131 700132
ILMV700131EA 0 INR InStock
ILMV700131 ILMV700132
Ball check valves, manual, ABV series
Valves Vacuum Valves
These ball check valves have a PTFE hollow ball and viton seals.
  • Compact design
  • Housing made of aluminium
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms