Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
VWR® UltraPure, PBS and TBS wash buffers
Supplier: VWR Chemicals
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
0788-2PKEA 0 INR InStock
0788-2PK E703-1L E703-500ML J640-4L
VWR® UltraPure, PBS and TBS wash buffers
Buffers Electrophoresis Buffers Washing Buffers
Wash buffers ideal for use in applications such as Western blotting and ELISA. Prevents high background by effectively removing unbound reagents.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms