Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
VWR® UltraPure, Buffer, PBS solution (phosphate buffered saline)
Supplier: VWR Chemicals
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
K812-20LEA 0 INR InStock
K812-20L
VWR® UltraPure, Buffer, PBS solution (phosphate buffered saline)
Buffers Cell Culture Buffers
Dulbecco's formulation without calcium and magnesium.
  • Free from DNase, RNase or protease
137,7 mM sodium chloride, 2,7 mM potassium chloride, 9,55 mM phosphate buffer
pH @ 25 °C      7,1 to 7,7
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms