Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
Wipe rolls, WYPALL* L40
Supplier: KIMBERLY CLARK
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
05701
KIMB05701EA 0 INR InStock
KIMB05701
Wipe rolls, WYPALL* L40
Wipes Disposable Wipes and Towels Wipes for General Applications
These single ply wipes are made from AIRFLEX* material, a specialised fabric with a perfect balance of softness and strength, that is also thick and absorbent. Ideal for industrial tasks such as cleaning, process line wiping, soaking up large spills.
  • Made with double re-creped (DRC) base sheet technology
  • Large, perforated rolls for use in busy areas
  • Also suitable for use in healthcare for wiping patients’ hands and face
  • Can be used with a dispensing system to help control use and reduce wastage
Matching dispensers: 115-2038, 115-0045 and 115-1533.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms