Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Nonidet® P 40 Substitute (NP-40), Proteomics Grade

Supplier: VWR Chemicals

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Warning

M158-100MLEA 0 INR InStock
M158-100ML
Nonidet® P 40 Substitute (NP-40), Proteomics Grade
Nonidet® P 40 Substitute (NP-40)
Non ionic detergent efficiently separates hydrophilic proteins from membrane spanning, hydrophobic proteins without altering biological activity.

  • High purity and protease-free
  • Suitable for most biological applications

Non-ionic detergents have uncharged hydrophilic groups and are gentle, non-denaturing detergents ideal for studying the conformation and function of proteins, because they keep secondary and tertiary structure intact.


Boiling Pt: 290 °C (1013 hPa)
Melting Pt: 57…58 °C
Density: 1.06 g/cm³ (20 °C)
Flash Pt: 113 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00132851
CAS Number: 9016-45-9
EINECS: 500-024-6
UN: 3082
ADR: 9 III
Order Now
Specification Test Results
Density (20 °C) 1.05 - 1.07 g/ml
Protease NONE
Refractive Index 1.48 - 1.52
Solubility (5%, Water) PASS

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

WRITE A REVIEW

REVIEWS (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

Print These Terms