Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
Macron Fine Chemicals™ StandARd®, Platinum Cobalt Colour Standard (APHA 500)
Supplier: MACRON AVANTOR BRAND
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
H296-03EA 0 INR InStock
H296-03 MACRH296-01
Macron Fine Chemicals™ StandARd®, Platinum Cobalt Colour Standard (APHA 500)
Standards Colour Reference and Spectroscopy Standards
Corrosive.
APHA 500 contains 0,13% chloroplatinic acid, 0,11% cobalt (II) chloride, and 4,6% hydrogen chloride in water.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms