Water, Molecular biology grade_meetSpec_USP Molecular biology grade, sterile, Corning®

Supplier: MEDIATECH
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

46-000-CV
MDTC46-000-CVEA 33948.62 INR
MDTC46-000-CV
Water, Molecular biology grade_meetSpec_USP Molecular biology grade, sterile, Corning®
Water

Molecular biology grade water is ideal for a wide variety of routine tasks requiring a high degree of sterility Molecular biology grade water is ideal for the preparation of reagents, rinsing glassware and plasticware, and other molecular biology applications where RNase-, DNase-, and Protease-free water is required. No toxic agents, including DEPC, are used in the preparation of Mediatech's molecular biology grade water, eliminating possible interferences with enzymatic reactions.


Certifications: Molecular Biology Grade Water Tested to USP Sterile Purified Water Specifications

Formula: H₂O
MW: 18.02 g/mol
Boiling Pt: 100
Melting Pt: 0
MDL Number: MFCD00011332
CAS Number: 7732-18-5
Merck Index: 13,10098

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR