Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
Garment racks, upright and wall mounted
Supplier: METRO
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
GRU2436S GRU2448S GRU2460S GRU2472S
METOGRU2436SEA 0 INR InStock
METOGRU2436S METOGRU2448S METOGRU2460S METOGRU2472S
Garment racks, upright and wall mounted
Furniture Gowning Racks
Racks are designed to provide efficient storage of cleanroom garments. The hangers are spaced at 75 mm intervals.
  • Stainless steel 304 with electropolished finish
  • Open-wire design promotes laminar air flow
  • Adjustable stainless steel levelling feet provides floor mount or free standing option
  • Unique hanger capture mechanism and hanger design provide theft deterrence
ESD classification: non-conductive.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms