Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
Utility Carts, MetroMax i®, Metro
Supplier: METRO
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
MXUC1830G-25 MXUC2436G-25
METOMXUC1830G-25EA 0 INR InStock
METOMXUC1830G-25 METOMXUC2436G-25
Utility Carts, MetroMax i®, Metro
Trolleys
Utility carts can be used in a variety of transport applications. The ergonomic handles and premium castors promote long life and ease of use. Microban product protection is built into the shelf mats, frames and posts. They are easy to clean, with removable shelf mats.
Corrosion proof shelves, posts and stainless steel 304 handle
Weight capacity: 408 kg.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms