Masterflex® Inert In-Line Check Valves, PEEK, Avantor®
Supplier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
01355-00 01355-04 01355-12 01355-16 01355-24 01355-28
MFLX01355-00EA 12226.42 INR
MFLX01355-00 MFLX01355-04 MFLX01355-12 MFLX01355-16 MFLX01355-24 MFLX01355-28
Masterflex® Inert In-Line Check Valves, PEEK, Avantor®
Valves Check Valves
No maintenance required!
  • Ideal for high-purity, low pressure
  • No metal components - inert flow path
  • No back flow leakage with back pressure of 0 to 100 psi
  • 1/4-28 UNF threaded connections
Unlike spring-actuated check valves that may restrict or impede the flow path, this in-line check valve features a smooth flow path that minimizes shear and turbulence.