Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
Masterflex® Manifold Mixing Solenoid Valves, Avantor®
Supplier: Avantor Fluid Handling
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
01356-00 01356-02 01356-03 01356-07 01356-08 01356-10 01356-14 01356-20 01356-22 01356-01 01356-04 01356-05 01356-09 01356-11 01356-12 01356-16 01356-18 01356-19 01356-21 01356-23
MFLX01356-00EA 63800 INR InStock
MFLX01356-00 MFLX01356-02 MFLX01356-03 MFLX01356-07 MFLX01356-08 MFLX01356-10 MFLX01356-14 MFLX01356-20 MFLX01356-22 MFLX01356-01 MFLX01356-04 MFLX01356-05 MFLX01356-09 MFLX01356-11 MFLX01356-12 MFLX01356-16 MFLX01356-18 MFLX01356-19 MFLX01356-21 MFLX01356-23
Masterflex® Manifold Mixing Solenoid Valves, Avantor®
Valves Solenoid Valves
Stand up to aggressive fluids.
  • Excellent for multiple liquid or gas control systems with aggressive or high-purity fluids
  • PTFE or PEEK wetted parts
  • Continuous duty and high cycle life
  • Suitable for pressure or vacuum service
These normally closed solenoid mixing valves feature low power consumption, minimal dead volume, and fast response time for all your mixing needs. All valves come with 15" (26 gauge) PTFE coated lead wires. Special orders are available for valves with different diaphragm and body material, or larger flow values — call for moreMore Product Information
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms