Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
PVC Hose and Cuffs
Supplier: Avantor Fluid Handling
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
06308-40
MFLX06308-40EA 55300 INR InStock
MFLX06308-40
PVC Hose and Cuffs
Tubing Transfer Tubing
This PVC hose is ideal for transfer, vacuum, and high flow rate applications. Heavy-duty, flexible construction resists moisture and chemical corrosion. "Spring-back" coil provides optimum crush resistance. maximum operating pressure is 44 psi at 72 °F; temperature range is 30 to 180 °F (-1 to 82 °C). Order PVC cuffs for simple, air tight hose installation.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms