Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
Parker Hannifin Valves Manual and Actuated Accessories
Supplier: Avantor Fluid Handling
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
06394-08
MFLX06394-08EA 2600 INR InStock
MFLX06394-08
Parker Hannifin Valves Manual and Actuated Accessories
Valves Needle Valves
Panel mount your Parker needle valve with ease.
  • Constructed of durable stainless steel
Choose from three panel nut options for 1/4", 3/8", or 1/2" Parker needle valves.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms