Masterflex® L/S® High-Performance Precision Pump Tubing, Versilon™ A-60-N, Avantor®
Supplier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
MFLX06404-15EA 13991.84 INR
MFLX06404-15 MFLX06404-24 MFLX06404-35 MFLX06404-36
Masterflex® L/S® High-Performance Precision Pump Tubing, Versilon™ A-60-N, Avantor®
Tubing Peristaltic Pump Tubing
Well-suited for vacuum/pressure applications.
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include packs, cases, and spools for customer-
More Product Information