Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
Chemical-Duty Self-Priming Transfer Pumps
Supplier: Avantor Fluid Handling
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
07190-70
MFLX07190-70EA 429000 INR InStock
MFLX07190-70
Chemical-Duty Self-Priming Transfer Pumps
Pumps Drum/Transfer Pumps
Different wetted materials for different chemical compatibilities.
  • Pumps with glass-filled polypropylene heads are ideal for transferring a wide range of caustic materials
  • Pumps with virgin PVDF heads are ideal for DI H2O, strong acids, and halogens
Self-priming pumps with glass-filled polypropylene heads are capable of lifting water up to 10 feet.
Delivery information: Includes conduit box for hardwiring.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms