LinkTech 20AC & 20PP Quick-Disconnect Fitting Insert, Compression without Valve
Supplier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
MFLX16360-37EA 17000 INR
MFLX16360-37
LinkTech 20AC & 20PP Quick-Disconnect Fitting Insert, Compression without Valve
Fittings and Connectors
Simply press the stainless steel thumb latch to disconnect the fittings.
  • Audible click lets you know the coupling is secure
  • Interchangeable with other similar industry thumb-latch fittings
Use these fittings to quickly connect and disconnect from your flow line. All fittings have 1/8" flow size, allowing you to mix and match between the fittings—simply order a body (female half) and an insert (male half). Couplings are compatible for use with gases and liquids. Choose between acetal for a wide operating temperature range and greater durability orMore Product Information