Labnet Spectrafuge™ 24D Digital Microcentrifuge
Supplier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
17415-10 17415-40 17410-45 17415-00 17415-15 17415-30
MFLX17415-10EA 0 INR
MFLX17415-10 MFLX17415-40 MFLX17410-45 MFLX17415-00 MFLX17415-15 MFLX17415-30
Labnet Spectrafuge™ 24D Digital Microcentrifuge
Centrifuges Microcentrifuges
Precise digital control, high capacity, and a maintenance-free brushless drive.
  • Exceptionally quite and cool running
  • Large display of rpm or g-force
  • Unique 24x1.5/2.0mL easy-access rotor
  • Pick your favorite color!
The digital microprocessor precisely regulates all aspects of operation. Run times from 30 seconds to 30 minutes, or continuously are easily set with the timer knob. Press and hold the knob for a quick spin — elapsed time is shown on the display.
Delivery information: Includes instruction manual, warranty card, power cord, standard rotor,More Product Information