Replacement adapters for IKA mini G centrifuges
Supplier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
17701-04
MFLX17701-04EA 0 INR
MFLX17701-04
Replacement adapters for IKA mini G centrifuges
Centrifuges Rotors
Use adapters to accommodate 0.5, 0.4 and 0.2 ml tubes.
Choose from a wide range of replacement adapters for your IKA mini G centrifuge.