Thermo Scientific Pico and Fresco Microcentrifuges
Supplier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
17703-05 17703-00 17703-10 17703-15 17703-45 17703-55
MFLX17703-05EA 0 INR
MFLX17703-05 MFLX17703-00 MFLX17703-10 MFLX17703-15 MFLX17703-45 MFLX17703-55
Thermo Scientific Pico and Fresco Microcentrifuges
Centrifuges Microcentrifuges
Powerful centrifugation in a small footprint.
  • Generous sample capacity in a quiet compact unit
These high-speed, compact microcentrifuges provide efficient sample processing and a generous sample capacity. Quiet motor with fast acceleration and deceleration allows you to minimize noise and processing time. Units feature a sleek, lightweight design for easy clean-up and minimal benchtop utilization. LED controls provide easy set-up and operation.