Process Cartridge Filter Housings, Stainless Steel
Supplier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
29808-02
MFLX29808-02EA 575000 INR
MFLX29808-02
Process Cartridge Filter Housings, Stainless Steel
Filters Filter Cartridges
Combination cartridge filter and centrifugal separator extends cartridge life and reduces maintenance.
  • Up-flow design with tangential entry prevents air entrapment
  • Process flow rates up to 150 GPM (568 LPM)
  • Electropolished 304 stainless steel housing with secure closure is NSF 61 listed
The unique design of these housings incorporates two filtration technologies in one. Fluid enters a tangential inlet into an outer chamber where centrifugal separation of heavier solids takes place. The fluid is then directed into the inner chamber for filtrationMore Product Information