Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
Masterflex® Gear Pump Systems, Low Flow, Avantor®
Supplier: Avantor Fluid Handling
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
70780-09
MFLX70780-09EA 1722000 INR InStock
MFLX70780-09
Masterflex® Gear Pump Systems, Low Flow, Avantor®
Pumps Gear Pumps
Choose the pump system configured best for your needs.
  • Maximum flow from 0.0081 to 0.3 depending on model│Configured skid mounted on a steel base
This pulseless flow pump system can be configured in a variety of ways to accommodate flow rates from 0.0009 to 0.30 GPM.
Please contact your local Avantor office to customize a system suited to your needs.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms