Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
General-Purpose TEFC/ODP, NEMA Type C-Face Four-In-One Motors
Supplier: Avantor Fluid Handling
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
71004-25 71005-15 71005-30 71004-15 71004-35 71004-45 71005-35
MFLX71004-25EA 113900 INR InStock
MFLX71004-25 MFLX71005-15 MFLX71005-30 MFLX71004-15 MFLX71004-35 MFLX71004-45 MFLX71005-35
General-Purpose TEFC/ODP, NEMA Type C-Face Four-In-One Motors
Pumps Motors
Totally enclosed motors are ideal for dusty, dirty, or damp environments.
  • Unique four-in-one design allows for replacement of ODP, TEFC, C-Face, or rigid based motors
  • Use for vertical or horizontal applications
Single-phase motors include a removable, rigid base. Three-phase motors inlcude a removable base.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms