Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
ITT Jabsco High-Purity Air-Operated Double-Diaphragm Pumps
Supplier: Avantor Fluid Handling
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
74020-00 74020-10
MFLX74020-00EA 564000 INR InStock
MFLX74020-00 MFLX74020-10
ITT Jabsco High-Purity Air-Operated Double-Diaphragm Pumps
Pumps Air-Operated Pumps
Unique air valve ensures a positive shift every time – pump will not stall.
  • All diaphragms, liquid chambers, and manifolds are constructed
  • of 100% virgin PTFE for long life and high purity
  • Unique valve requires no lubrication or prepacking.
  • Easily access air valve without disassembling the pump
Pumps are manufactured under class 1000 clean room conditions to guarantee purity.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms