Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
Masterflex® L/S® Analog Variable-Speed Console Drive Systems, Avantor®
Supplier: Avantor Fluid Handling
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
MFLX77910-45-INEA 241000 INR InStock
MFLX77910-45-IN MFLX77910-55-IN MFLX77910-65-IN MFLX77910-75-IN MFLX77910-45-IL MFLX77910-55-IL MFLX77910-65-IL MFLX77910-75-IL MFLX77910-45-CH MFLX77910-55-CH MFLX77910-65-CH MFLX77910-75-CH
Masterflex® L/S® Analog Variable-Speed Console Drive Systems, Avantor®
Pumps Peristaltic Pumps
Compact, cost effective pump systems for general fluid transfer applications.
  • Simple operation with single-turn speed control
  • Separate power switch enables you to maintain speed setting when turning drive on/off; front-panel LED indicates power on
  • Reversible motor lets you easily prime/purge tubing and pump in either direction
  • Remote control of start/stop via DB9 connector on back of drive
Choose a convenient, complete system all with one catalog number.
Order Now
ORDER
SERVICES
RELATED INFORMATION
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms