Masterflex® Flow-Through Pump Tubing Weights, Avantor®
Supplier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
MFLX07570-04EA 18400 INR
MFLX07570-04 MFLX78226-81 MFLX78226-82
Masterflex® Flow-Through Pump Tubing Weights, Avantor®
Weights Sinkers
Keep your samples flowing and tubing from curling.
  • Choose between PTFE or stainless steel construction
This flow-through tubing weight prevents tubing from curling or floating. Ideal for groundwater sampling, this weight fits Masterflex®; L/S® 15 or L/S® 24 Precision tubing.