Masterflex® Ismatec® Pump Tubing, 3-Stop Color-Coded Autoanalysis/Microbore, Autoclavable PharMed® BPT, Avantor®
Supplier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
95693-48
MFLX95693-48EA 0 INR
MFLX95693-48
Masterflex® Ismatec® Pump Tubing, 3-Stop Color-Coded Autoanalysis/Microbore, Autoclavable PharMed® BPT, Avantor®
Tubing Peristaltic Pump Tubing
Designed for optimal operation in your Ismatec pumps.
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory approvals to match your unique application
This tubing features welded, not glued, tubing stops - no adhesive, so the tubing sets withstand autoclaving. Tubing set has 2 actual welded stops for use in a 3 stop pump. The overall lengthMore Product Information