Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
Clinical Chemistry Analyser, BS 240 Pro
Supplier: SHENZHEN MINDRAY BIO MEDICAL
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
BS-240PRO
MINDBS-240PROEA 1297664.91 INR InStock
MINDBS-240PRO
Clinical Chemistry Analyser, BS 240 Pro
Clinical Diagnostic Supplies Clinical Analysers
BS 240 Pro, a compact Robust Chemistry Analyzer.
  • Constant throughput of 240 photometric T/H
  • Large and Flexible capacity with upto 100 sample positions and upto 100 reagent positions (50 Fixed + 50 interchangeable)
  • Grating Photometer (100 ul minimum reaction volume)
  • Easy loading and unloading for samples; One key calibrator import
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms