Portable balances, Navigator® and Navigator® XL series
Supplier: OHAUS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
83033085
OHAU83033085EA 0 INR
OHAU83033085
Portable balances, Navigator® and Navigator® XL series
Balances and Scales
Two series of lightweight, robust, portable balances that are ideal for use in a wide range of laboratory and industrial applications. The innovative, touch-free sensors allow up to 36 combinations of functions including tare, zero, print, function and, where appropriate, display backlight. The applications include weighing (with 14 units), percentage weighing, check weighing, parts counting, accumulation/totalisation and display hold. The calibration function is digital and requires an external weight.
  • Dual multi-functional touch-free sensors free up your hands, keeping the balance
More Product Information