Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
Analytical sieve shakers, AS 200/AS 300
Supplier: RETSCH
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
30.030.0001 30.032.0001
RETS30.030.0001EA 0 INR InStock
RETS30.030.0001 RETS30.032.0001
Analytical sieve shakers, AS 200/AS 300
Shakers Sieve Shakers
Retsch analytical sieve shakers are used in research and development, quality control of raw materials, interim and finished products, as well as in production monitoring.
  • Sieving with 3-D effect
  • For wet and dry sieving
  • Short sieving times
  • Low noise, maintenance-free
AS 200 basic; favourably priced basic model.
AS 200 digit cA; powerful comfort model with touch-sensitive keypad, digital display and interval sieving.
AS 200 control; as AS 200 digit, but microprocessor controlled, with freely selectable intervals and digital vibration amplitudeMore Product Information
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms