Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
Serum replacement, FreeAdd 1X
Supplier: BIOWEST
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
S6010-050 S6010-500
S6010-050EA 0 INR InStock
S6010-050 S6010-500
Serum replacement, FreeAdd 1X
Sera (Serum)
FreeAdd 1X is a fully chemically defined substitute for animal serum. It provides the necessary nutritional support for cell growth, development and expression. It has been tested on a variety of cells and can be used in combination with basal medium to generate equal or even better cell culture performance.
  • No hydrolysates or mixtures
  • Non detectable endotoxin level
  • Broad range (cell lines, primary cells, stem cells)
  • GMP produced
  • 100% batch-to-batch consistency
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms