Calibration weights, class E2, Proof Line
Supplier: Sartorius Balances
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
YCW0121-02 YCW0221-02 YCW312-02 YCW512-02 YCW422-02 YCW612-00 YCW622-00 YCW452-02 YCW522-00 YCW612-02 YCW622-02 YCW412-00 YCW322-02 YCW352-00 YCW712-00 YCW1221-00 YCW2521-00 YCW2221-02 YCW1521-02 YCW0121-00 YCW2521-02 YCW1521-00 YCW2121-00 YCW2121-02 YCW2221-00 YCW0221-00 YCW412-02 YCW652-02 YCW422-00 YCW712-02 YCW352-02 YCW552-02 YCW312-00 YCW512-00 YCW322-00 YCW552-00 YCW652-00 YCW1121-00 YCW0521-00 YCW1221-AC-00 YCW312-AC-00 YCW2521-AC-02 YCW2521-AC-00 YCW2221-AC-02 YCW2221-AC-00 YCW2121-AC-02 YCW2121-AC-00 YCW1521-AC-02 YCW1221-AC-02 YCW1121-AC-02 YCW1121-AC-00 YCW0521-AC-02 YCW0521-AC-00 YCW0221-AC-02 YCW0221-AC-00 YCW0121-AC-02 YCW0121-AC-00 YCW1521-AC-00 YCW352-AC-02 YCW352-AC-00 YCW312-AC-02 YCW322-AC-02 YCW322-AC-00 YCW412-AC-00 YCW622-AC-02 YCW622-AC-00 YCW612-AC-02 YCW612-AC-00 YCW652-AC-00 YCW652-AC-02 YCW712-AC-02 YCW712-AC-00 YCW452-AC-02 YCW452-AC-00 YCW422-AC-02 YCW422-AC-00 YCW412-AC-02 YCW522-AC-00 YCW552-AC-02 YCW552-AC-00 YCW522-AC-02 YCW512-AC-02 YCW512-AC-00
SARBYCW0121-02EA 0 INR
SARBYCW0121-02 SARBYCW0221-02 SARBYCW312-02 SARBYCW512-02 SARBYCW422-02 SARBYCW612-00 SARBYCW622-00 SARBYCW452-02 611-1705 SARBYCW612-02 SARBYCW622-02 SARBYCW412-00 SARBYCW322-02 SARBYCW352-00 SARBYCW712-00 SARBYCW1221-00 SARBYCW2521-00 SARBYCW2221-02 SARBYCW1521-02 SARBYCW0121-00 SARBYCW2521-02 SARBYCW1521-00 SARBYCW2121-00 SARBYCW2121-02 SARBYCW2221-00 SARBYCW0221-00 SARBYCW412-02 SARBYCW652-02 SARBYCW422-00 SARBYCW712-02 SARBYCW352-02 SARBYCW552-02 SARBYCW312-00 611-1704 SARBYCW322-00 611-1706 611-1709 SARBYCW1121-00 SARBYCW0521-00 SARBYCW1221-AC-00 SARBYCW312-AC-00 SARBYCW2521-AC-02 SARBYCW2521-AC-00 SARBYCW2221-AC-02 SARBYCW2221-AC-00 SARBYCW2121-AC-02 SARBYCW2121-AC-00 SARBYCW1521-AC-02 SARBYCW1221-AC-02 SARBYCW1121-AC-02 SARBYCW1121-AC-00 SARBYCW0521-AC-02 SARBYCW0521-AC-00 SARBYCW0221-AC-02 SARBYCW0221-AC-00 SARBYCW0121-AC-02 SARBYCW0121-AC-00 SARBYCW1521-AC-00 SARBYCW352-AC-02 SARBYCW352-AC-00 SARBYCW312-AC-02 SARBYCW322-AC-02 SARBYCW322-AC-00 SARBYCW412-AC-00 SARBYCW622-AC-02 SARBYCW622-AC-00 SARBYCW612-AC-02 SARBYCW612-AC-00 SARBYCW652-AC-00 SARBYCW652-AC-02 SARBYCW712-AC-02 SARBYCW712-AC-00 SARBYCW452-AC-02 SARBYCW452-AC-00 SARBYCW422-AC-02 SARBYCW422-AC-00 SARBYCW412-AC-02 SARBYCW522-AC-00 SARBYCW552-AC-02 SARBYCW552-AC-00 SARBYCW522-AC-02 SARBYCW512-AC-02 SARBYCW512-AC-00
Calibration weights, class E2, Proof Line
Weights Calibration Weights
Sartorius Proof Line weights are high quality working standards for everyday use. These weights are made of high quality material with polished or precision lathed surfaces.
  • 0 to 5 mg leaf weights: Aluminium
  • 10 to 500 mg leaf weights: Nickel silver
  • 1 g to 10 Kg Knob weights: Stainless steel
Certifications: Comply in all respects with the requirements on shape, material and markings specified by the OIML R111-1 International Recommendation.