Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
Flasks, flat bottom, with standard ground joints, DURAN®
Supplier: DWK LIFE SCIENCES
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
241714408 241716306 241712407 241712604 241712707 241713609 241713703 241714605 241715404 241715601
SCOT241714408EA 0 INR InStock
SCOT241714408 SCOT241716306 SCOT241712407 SCOT241712604 SCOT241712707 SCOT241713609 SCOT241713703 SCOT241714605 SCOT241715404 SCOT241715601
Flasks, flat bottom, with standard ground joints, DURAN®
Flasks Flat Bottom Flasks
DURAN®, borosilicate glass 3,3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
With labelling area.
*non-DIN ISO size
Certifications: According to DIN ISO 4797
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms