Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
Screwthread tubes, attachable, DIN thread, DURAN®
Supplier: DWK LIFE SCIENCES
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
248350103 248380209 248360207 248370105 248390107
SCOT248350103EA 0 INR InStock
SCOT248350103 SCOT248380209 SCOT248360207 SCOT248370105 SCOT248390107
Screwthread tubes, attachable, DIN thread, DURAN®
Adapters Adapters, Screwthread tubes
Made of DURAN®, borosilicate glass 3.3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Screw thread GL. It is used by glassblowers in the construction of laboratory glassware.
DIN 12216
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms