Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024
Chemical resistant gloves, uvex rubiflex S NB40S
Supplier: UVEX
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
9890222
UVEX9890222EA 0 INR InStock
UVEX9890222
Chemical resistant gloves, uvex rubiflex S NB40S
Gloves
These fully coated NBR (nitrile butadiene rubber) gloves provide excellent resistance to many chemicals, acids, alkalis, mineral oils and solvents.
  • Very good mechanical characteristics, outstanding abrasion resistance
  • Good water vapour absorption due to the cotton lining
  • Anatomic shape, highly flexible
  • Reinforced design
Ideal for use in the automotive, chemical, mechanical industry as well as for metal processing.
Certifications: EN ISO 374:2016 Type A (JKNOPT), EN 388:2016 (2121X)
Food contact approved
OEKO-Tex® Standard 100
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms