Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

n-Heptane 99 GC, corrected for water content, UltimAR® LC_Organic_Residue_Analysis, UV, Macron Fine Chemicals™

Supplier: MACRON AVANTOR BRAND

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Danger

V554-10EA 0 INR InStock
V554-10
n-Heptane 99 GC, corrected for water content, UltimAR® LC_Organic_Residue_Analysis, UV, Macron Fine Chemicals™
n-Heptane
Suitable for liquid chromatography, extract/conc, UV-spectrophotometry, organic residue analysis.


Formula: H₃C(CH₂)₅CH₃
MW: 100.20 g/mol
Boiling Pt: 98.4
Melting Pt: –90.6 °C
Density: 0.6838 g/cm³ (20 °C)
Flash Pt: –4 °C
Storage Temperature: Ambient
MDL Number: MFCD00009544
CAS Number: 142-82-5
EINECS: 205-563-8
UN: 1206
ADR: 3 II
Merck Index: 13,04679
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8ff97126664c92362bf0aa2645719548c99f0533&vid=8448ee44d4a93782bfa2900878099424dfa17053&prid=web&clid=SAPDAM
Order Now
Specification Test Results
For Laboratory,Research,or Manufacturing Use
ACS - Assay (CH₃(CH₂)₅CH₃) (by GC, corrected for water) ≥99.0 %
ACS - Color (APHA) ≤10
ACS - Residue after Evaporation ≤1 ppm
ACS - Substances Darkened by H₂SO₄ Passes Test
ACS - Water (H₂O)(by KF, coulometric) ≤0.01 %
Ultraviolet Absorbance (1.00-cm cell vs. water) - 197 nm ≤1.00
Ultraviolet Absorbance (1.00-cm cell vs. water) - 210 nm ≤0.40
Ultraviolet Absorbance (1.00-cm cell vs. water) - 220 nm ≤0.10
Ultraviolet Absorbance (1.00-cm cell vs. water) - 254 - 400 nm ≤0.01
Fluorescence Trace Impurities, measured as Quinine Base - at Emission Maximum for Impurities ≤1.0 ppb
ECD Sensitive Impurities (as Heptachl or Epoxide) Single Peak (ng/L) ≤10 ppb
FID Sensitive Impurities (as 2-Octano l) Single Peak (µg/L) ≤5 ppb
ACS - Neat Solvent Front Characterization Passes Test
Titrable Acid (µeq/g) ≤0.8

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

WRITE A REVIEW

REVIEWS (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

Print These Terms